missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ TNF alpha Monoclonal Antibody (4F4)
GREENER_CHOICE

Artikelnummer. 11838721
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 μg
Pakningsstørrelse:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11838721 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11838721 Leverandør Invitrogen™ Leverandørnr. MP390

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

MP390 detects natural and recombinant TNF alpha, with a predicted molecular weight of approximately 25 kDa. This antibody will not cross-react with human or mouse TNFa, mouse TNFb, or porcine TNFb. This is a carrier-free low endotoxin formulation, <1.0 EU/mg. This product can be used as a coating antibody in a sandwich ELISA (100 μg/well) in combination with biotin-labeled TNFa monoclonal antibody (Product # M302B, 0.1-0.65 μg/mL). For Western blot, this antibody has been successfully used in conjunction with 0.02 μg/mL Biotin-conjugated Goat-anti-Mouse IgG (Product # 31800), 0.05 μg/mL HRP-conjugated Streptavidin (Product # N100), and recombinant Porcine TNFa (Product # RPTNFAI), followed by visualization using SuperSignal™ West Pico Chemiluminescent Substrate (Product # 34080). This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).

TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen TNF alpha
Applikationer ELISA, Western Blot
Klassifikation Monoclonal
Klon 4F4
Koncentration 1 mg/mL
Konjugeret Unconjugated
Formulering PBS with no preservative
Gene Tnf
Genadgangsnr. P23563
Gene Alias APC1 protein; Cachectin; C-domain 1; C-domain 2; cTNF; DADB-70P7.1; DIF; ICD1; ICD2; Intracellular domain 1; Intracellular domain 2; N-terminal fragment; NTF; RATTNF; Tnf; TNF alpha; TNF superfamily; TNF α; TNF, macrophage-derived; TNF, monocyte-derived; TNFA; TNF-a; TNFalpha; TNF-alpha; Tnfsf1a; TNFSF2; TNFα; TNLG1F; Tumor necrosis factor; tumor necrosis factor (TNF superfamily, member 2); tumor necrosis factor alpha; tumor necrosis factor alpha (cachetin); tumor necrosis factor alpha precursor; tumor necrosis factor ligand 1F; tumor necrosis factor ligand superfamily member 2; Tumor necrosis factor, membrane form; Tumor necrosis factor, soluble form; tumor necrosis factor-alpha; tumor necrosis factor-alpha precursor; tumor-necrosis factor; tumour necrosis factor
Gensymboler Tnf
Værtsarter Mouse
Immunogen Recombinant porcine TNF alpha (Baculovirus-derived).
Oprensningsmetode Protein A
Mængde 500 μg
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 397086
Målarter Pig
Indhold og opbevaring -20°C
Produkttype Antibody
Form Liquid
Isotype IgG1
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.