missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
TMPRSS7 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-83702-0.1ML
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
TMPRSS7 Polyclonal specifically detects TMPRSS7 in Human samples. It is validated for Western Blot.
Tekniske data
| TMPRSS7 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 344805 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| transmembrane protease serine 7, transmembrane protease, serine 7, type II transmembrane serine protease 7 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human TMPRSS7 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion