Få mere at vide
TMEM41B Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-12857FR
Produktkode 18310887
  • 5135.00 DKK / 0.10ml
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
TMEM41B Polyclonal antibody specifically detects TMEM41B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY
Tekniske data
Tekniske data
TMEM41B
Polyclonal
50mM Sodium Borate with 0.05% Sodium Azide
Rabbit
Affinity-purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
DyLight 680
KIAA0033MGC33897, MGC57262, transmembrane protein 41B
Anti-TMEM41B antibody was raised against a peptide corresponding to 10 amino acids near the carboxyl terminus of human TMEM41B. The immunogen is located within the last 50 amino acids of TMEM41B.
0.1 mL
Autophagy, Lipid and Metabolism
440026
Store at 4°C in the dark.
IgG