missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
TMEM206 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
3770.00 DKK
Tekniske data
| Antigen | TMEM206 |
|---|---|
| Applikationer | Western Blot |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
| Værtsarter | Rabbit |
Beskrivelse
TMEM206 Polyclonal specifically detects TMEM206 in Human samples. It is validated for Western Blot.Tekniske data
| TMEM206 | |
| Polyclonal | |
| Rabbit | |
| transmembrane protein 206 | |
| TMEM206 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 55248 | |
| Synthetic peptides corresponding to C1ORF75 The peptide sequence was selected from the N terminal of C1ORF75. Peptide sequence IFIYLLLMAVAVFLVYRTITDFREKLKHPVMSVSYKEVDRYDAPGIALYP. | |
| Primary |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel