missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM119 Mouse anti-Human, Unconjugated, Clone: 1E7A2, Proteintech
Mouse Monoclonal Antibody
1043.29 DKK - 2973.37 DKK
Specifications
Antigen | TMEM119 |
---|---|
Clone | 1E7A2 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16869265
|
Proteintech
66948-1-IG-20UL |
20 μL |
1043.29 DKK
20µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16859265
|
Proteintech
66948-1-IG-150UL |
150 μL |
2973.37 DKK
150µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The TMEM119 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TMEM119. This antibody recognizes human, pig, mouse, rat antigen. The TMEM119 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
TMEM119 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
338773 | |
Primary |
1E7A2 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human, Mouse, Rat, Pig | |
IgG3 |