missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMCO5 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-06174V
Additional Details : Weight : 0.00970kg
Description
TMCO5 Polyclonal specifically detects TMCO5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TMCO5 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
TMCO5, Transmembrane And Coiled-Coil Domain-Containing Protein 5A, Transmembrane And Coiled-Coil Domains 5, Transmembrane And Coiled-Coil Domains 5A | |
Produced in rabbits immunized with E. coli-derived Human TMCO5 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
145942 | |
Store at 4°C in the dark. | |
IgG |