Learn More
Abcam TMB ELISA Substrate (Slower Kinetic Rate), 1000 mL
Product Code. 32108454 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
Product Code. Quantity Unit Size
32108454 1000 mL 1000mL
1 options
Product Code. 32108454 Supplier Abcam Supplier No. ab1715261000mL
 Restricted Item
Forventet forsendelse: 03-12-2026
Abcam TMB ELISA Substrate (Slower Kinetic Rate).
  • Species Reactivity: Species independent
  • Storage: Store in the dark
Specifications
Product Type ELISA Substrate
Quantity 1000 mL
For Use With (Application) ELISA
Content And Storage Store in the dark