Få mere at vide
Novus Biologicals™ TM4SF1/L6 Antibody (Beth Israel Patent Anti-TM4SF1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28479-100ug
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
TM4SF1/L6 Monoclonal antibody specifically detects TM4SF1/L6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Tekniske data
| TM4SF1/L6 | |
| Monoclonal | |
| Unconjugated | |
| P30408 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Beth Israel Patent Anti-TM4SF1 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| L6, M3S1Tumor-associated antigen L6, Membrane component chromosome 3 surface marker 1, membrane component, chromosome 3, surface marker 1, TAAL6H-L6, transmembrane 4 L six family member 1, transmembrane 4 L6 family member 1, transmembrane 4 superfamily member 1 | |
| TM4SF1 | |
| 100 μg | |
| Cancer, Cell Cycle and Replication | |
| 4071 | |
| Human | |
| Purified |