missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
TLR2 Antibody (TL2.1), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-12062V
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
TLR2 Monoclonal antibody specifically detects TLR2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
Tekniske data
| TLR2 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| IgG κ |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| TL2.1 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
| This antibody was raised by immunising BALB/c mice with CHO cells transfected with human TLR2 cDNA. | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
| 7097 | |
| Store at 4°C in the dark. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion