missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
TLR12 Antibody (1229C) [Janelia Fluor™ 646], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB8086J
This item is not returnable.
View return policy
Description
TLR12 Monoclonal antibody specifically detects TLR12 in Mouse samples. It is validated for Western Blot, Flow Cytometry
Specifications
| TLR12 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 100131451 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry | |
| 1229C | |
| Western Blot, Flow Cytometry | |
| toll-like receptor 12 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse TLR12, Thr22-Lys709, Accession # Q6QNU9 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction