missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TLK2 Recombinant Human Protein

TLK2 or Tousled-like kinase 2, is a nuclear serine/threonine kinases. Recombinant human TLK2 (388-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.

Brand:  Invitrogen™ PV6426

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15479043

  • 55700.00 DKK / 1mg
Estimated Shipment: 14-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TLK2 is thought to be involved in the regulation of chromatin assembly. In addition, TLK2 is also involved in other cellular processes such as intracellular signaling cascade, protein amino acid phosphorylation, cell cycle, chromatin modification and in response to DNA damage stimulus. TLK2 displays maximal activity during S phase of the cell cycle where it appears to be regulated by cell-cycle-dependent phosphorylation. Inhibition of DNA replication causes a rapid loss of TLK2 activity, indicating that TLK2 function is tightly linked to ongoing DNA replication.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications

Specifications

Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
68.2kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
aa 388-end
Recombinant
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol
1mg
MGC44450, PKU-ALPHA
Human
GST Tag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers