Få mere at vide
Tocris Bioscience™ TL 13-22
Beskrivelse
Negative control for TL 13-12 (Cat. No. 6744). Displays no degradation of ALK in cell lines. Highly potent inhibitor of ALK (IC50 = 0.54 nM)
TL 13-22 is a negative control for TL 13-12 (Cat. No. 6744). Displays no degradation of ALK in cell lines. Highly potent inhibitor of ALK (IC50 = 0.54 nM)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-(2-(2-(2-(4-(4-((5-Chloro-4-((2-(isopropylsulfonyl)phenyl)amino)pyrimidin-2-yl)amino)-3-methoxyphenyl)piperazin-1-yl)ethoxy)ethoxy)ethyl)-2-((1,3-dioxo-2-(2-oxopiperidin-3-yl)isoindolin-4-yl)amino)acetamide |
| CAS | 2229036-65-9 |
| Mængde | 5 mg |
| Molekylær formel | C45H55ClN10O9S |
| Formel vægt | 947.5 |
| Renhed | 98% |