Få mere at vide
Tivozanib, TRC
Artikelnummer. 30132565
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132565 50 mg
30011499 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30132565 Leverandør Toronto Research Chemicals Leverandørnr. TRCT44720550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 475108-18-0
Molekylær formel C22 H19 Cl N4 O5
Molekylvægt (g/mol) 454.86
Synonym Urea, N-[2-chloro-4-[(6,7-dimethoxy-4-quinolinyl)oxy]phenyl]-N'-(5-methyl-3-isoxazolyl)-, N-[2-Chloro-4-[(6,7-dimethoxy-4-quinolinyl)oxy]phenyl]-N'-(5-methyl-3-isoxazolyl)urea, AV 951, KRN 951, Kil 8951, N-[2-Chloro-4-[(6,7-dimethoxy-4-quinolyl)oxy]phenyl]-N'-(5-methyl-3-isoxazolyl)urea, Tivozanib
IUPAC navn 1-[2-chloro-4-(6,7-dimethoxyquinolin-4-yl)oxyphenyl]-3-(5-methyl-1,2-oxazol-3-yl)urea
SMIL COc1cc2nccc(Oc3ccc(NC(=O)Nc4cc(C)on4)c(Cl)c3)c2cc1OC
Tekniske data
Kemisk navn eller materiale Tivozanib
Mængde 50 mg
InChI formel InChI=1S/C22H19ClN4O5/c1-12-8-21(27-32-12)26-22(28)25-16-5-4-13(9-15(16)23)31-18-6-7-24-17-11-20(30-3)19(29-2)10-14(17)18/h4-11H,1-3H3,(H2,25,26,27,28)
Formel vægt 454.1044