Learn More
RETSCH Titanium-niob Coated Knife
Brand:  RETSCH 02.446.0052
Additional Details : Weight : 0.21000kg
Product Code. 11914312
  • 7130.00 DKK / Each
Estimated Shipment: 02-08-2024
to see stock.
Specifications
Knife
For grinding without heavy-metal contamination
For GRINDOMIX GM 300
Titanium