Learn More
RETSCH Titanium-niob Coated Knife
Brand: RETSCH 02.446.0052
Additional Details : Weight : 0.21000kg
Specifications
Knife | |
For grinding without heavy-metal contamination | |
For GRINDOMIX GM 300 | |
Titanium |