Få mere at vide
Scharlab TISAB IV, for fluorides determination, accordingto ASTM D1179, 500 mL, MA: TI03300500, MSDS:
Brand:  Scharlab TI03300500
TISAB IV, for fluorides determination, accordingto ASTM D1179, 500 mL, MA: TI03300500, MSDS:
Produktkode 12472371
  • ONLINE EXCLUSIVE PRICE
    3144.75 DKK
    1911.17 DKK / 500ml
    Save 1233.58 DKK
    39% Slukket