Learn More
Tirasemtiv, MedChemExpress
Product Code. 16499477
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499477 5 mg 5mg
16437016 100 mg 100mg
16436938 10 mg 10mg
16412997 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16499477 Supplier Medchem Express Supplier No. HY15964
 Restricted Item
Forventet forsendelse: 08-12-2026
MedChemExpress Tirasemtiv is an activator of the fast skeletal muscle troponin complex.
Tirasemtiv is an activator of the fast skeletal muscle troponin complex.
Specifications
Chemical Name or Material Tirasemtiv
Physical Form Solid
CAS 1005491-05-3
Color Gray
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C12H14N4O
For Use With (Application) Neuroscience-Neuromodulation
Synonym CK-2017357
Solubility Information DMSO : 10.91 mg/mL (47.38 mM; Need ultrasonic)
SMILES O=C1N(C(CC)CC)C2=NC(C#C)=CN=C2N1
Molecular Weight (g/mol) 230.27
Formula Weight 230.27
Percent Purity 99.83%
Grade Research