Få mere at vide
Novus Biologicals™ TIRAP (TLR2 and TLR4) Inhibitor Peptide Set
Artikelnummer. p-200087451 Shop alle Bio Techne produkter
Mængde:
5 mg
Artikelnummer. 18708203
Brand: Novus Biologicals™ NBP226245
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Applications: Flow Cytometry, In vitro assay
TLR2 / TLR4 Inhibitor Peptide: 1mg (lyophilized); sequence: DRQIKIWFQNRRMKWKKPGFLRDPWCKYQML (Inhibitor sequence: PGFLRDPWCKYQML); Molecular weight: 4097.92 Da ; Antennapedia Control Peptide: 1mg (lyophilized); sequence: DRQIKIWFQNRRMKWKK; Molecular weight: 2361 Da
TRUSTED_SUSTAINABILITY
For Research Use Only