Learn More
Sampling Systems™ Tip for Unit Dose Sleeve Sampler
Developed for customers requiring accurate unit dose samples in small volumes. Sampling Systems™ Tip for Unit Dose Sleeve Sampler is ideal for use with a wide range of free flowing powders and granules.
Brand: Sampling Systems™ 1500W-05
This item is not returnable.
View return policy
Description
- For sampling small volume powders and granules.
- Chemically resistant 316L stainless steel construction.
- Different sample sizes can be taken using the same sampler
- Tips for use with Type 1 or Type 2 Sampling Chambers
Specifications
Unit Dose Sleeve Sampler Tip | |
0.5 mL | |
41.5 mm |
Stainless Steel (316L) | |
6 mm |
Product Content Correction
Product Title
Sampling Systems™ Tip for Unit Dose Sleeve Sampler > Capacity: 0.5mL
Spot an opportunity for improvement?Share a Content Correction