Få mere at vide
Tiotropium-d3 Bromide, TRC
Artikelnummer. 30452044
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30452044 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452044 Leverandør Toronto Research Chemicals Leverandørnr. TRCT44485210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tiotropium N-methyl-d3 bromide
CAS 1127226-56-5
Molekylær formel C19 D3 H19 N O4 S2 . Br
Mængde 10 mg
Synonym 3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane','7-[(2-hydroxy-2,2-di-2-thienylacetyl)oxy]-9-methyl-9-(methyl-d3)-','bromide (1:1)','(1α,2β,4β,5α,7β)-, Tiotropium-d3 Bromide, Tiotropium-(N-methyl-d3) bromide, (1α,2β,4β,5α,7β)-7-[(2-Hydroxy-2,2-di-2-thienylacetyl)oxy]-9-methyl-9-(methyl-d3)-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide, Tiotropium-D3 (N-methyl-D3) bromide
InChI formel InChI=1S/C19H22NO4S2.BrH/c1-20(2)12-9-11(10-13(20)17-16(12)24-17)23-18(21)19(22,14-5-3-7-25-14)15-6-4-8-26-15;/h3-8,11-13,16-17,22H,9-10H2,1-2H3;1H/q+1;/p-1/t11-,12-,13+,16-,17+;/i1D3;/t11-,12-,13+,16-,17+,20?;
SMIL [Br-].[2H]C([2H])([2H])[N+]1(C)[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)C(O)(c4cccs4)c5cccs5
IUPAC navn [(1R,2R,4S,5S)-9-methyl-9-(trideuteriomethyl)-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonan-7-yl] 2-hydroxy-2,2-bis(2-thienyl)acetate bromide
Molekylvægt (g/mol) 475.435
Formel vægt 474.036