Learn More
Tioconazole, TRC
Product Code. 30450805
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450805 100 g -
30456657 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30450805 Supplier Toronto Research Chemicals Supplier No. TRCT444680100G
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 65899-73-2
Molecular Formula C16 H13 Cl3 N2 O S
Molecular Weight (g/mol) 387.71
Synonym Tioconazole, 1-[(2RS)-2-[(2-Chlorothiophen-3-yl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole
IUPAC Name 1-[2-[(2-chlorothiophen-3-yl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole
SMILES Clc1ccc(C(Cn2ccnc2)OCc3ccsc3Cl)c(Cl)c1
Specifications
Quantity 100 g
InChI Formula InChI=1S/C16H13Cl3N2OS/c17-12-1-2-13(14(18)7-12)15(8-21-5-4-20-10-21)22-9-11-3-6-23-16(11)19/h1-7,10,15H,8-9H2
Formula Weight 385.9814
Chemical Name or Material Tioconazole