missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ VARIO™ DPD Free Chlorine Powder Pack Reagent, 25mL
Formulated for use in Lovibond test kits, colorimeters and spectrophotometers. Newly redesigned foil packs are easy to handle, easy to open and pour
Brand: Lovibond™ 530113
Additional Details : Weight : 0.30000kg
Description
- Range: 0 to 5.00mg/L
- For use in our test kits, colorimeters and spectrophotometers
- US EPA compliant
- Pre-measured, unit dose reagents
- Easy to handle, easy to open and easy to pour
- Case of 1000
- Fast and easy use of VARIO Powder Packs has made them extremely popular for water testing applications in many countries throughout the world
- Provides more experienced users with a real alternative to existing measurement systems
- Produced to the same high quality standards that have made Tintometer’s tablet reagents so successful for several decades
- Parameters from aluminum and chlorine through to sulphate are just some of the well-known tests that are included in the VARIO Powder Pack range
Specifications
DPD Free Chlorine Powder Pack Reagent | |
Test kits, colorimeters and spectrophotometers | |
25 mL | |
Vario™ Powder Reagents |
Water Testing | |
Case of 1000 | |
0 to 5.00mg/L |