Learn More
Lovibond™ Tintometer™ Universal pH Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand: Lovibond™ 515441BT
Additional Details : Weight : 0.30000kg
Description
- Universal pH Tablets
- Pack of 250
Specifications
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
250 Tablets | |
Universal pH Tablets |