missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ Thymol Blue Solution
For use with Lovibond test kits, colorimeters and spectrophotometers
Brand: Lovibond™ 451653
Additional Details : Weight : 0.30000kg
Description
- Thymol Blue Solution
- pH indicator
- 25mL, 100mL, 250mL and 500mL
Specifications
pH Test | |
For use with Lovibond test kits, colorimeters and spectrophotometers | |
500 mL | |
Thymol Blue Solution |