Learn More
Lovibond™ Tintometer™ Sulphate Turbitity Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
467.00 DKK - 1055.00 DKK
Specifications
For Use With (Application) Photometry Tablet Reagents
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Type Sulphate Turbitity Tablets
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15381845
View Documents Promotion Details
Lovibond™
515451BT
250 Tablets
1055.00 DKK
Pack of 250
View alternative products
Save Up to 
»
Estimated Shipment: 17-06-2024
to see stock.
Add to basket
15371845
View Documents Promotion Details
Lovibond™
515450BT
100 Tablets
467.00 DKK
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
Description
Description
  • New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
  • With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
  • Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
  • Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
  • ‘BT’ is added to the end of the code to identify the new style of packaging
  • Simple, time-saving and practical
  • There are no safety risks if the tablets are used in line with the instructions supplied
  • Safety data sheets are available for all reagents
Specifications
Specifications
Photometry Tablet Reagents
Test kits, colorimeters and spectrophotometers
Sulphate Turbitity Tablets