Learn More
Lovibond™ Tintometer™ Potassium T Tablets, 0.7 to 12mg/L
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand:  Lovibond™ 515670BT
Additional Details : Weight : 0.50000kg
Product Code. 10211792
  • 685.00 DKK / Pack of 100
Estimated Shipment: 01-07-2024
to see stock.
Add to basket
Description
Description
  • Range: 0.7 to 12mg/L
  • Parameter: Potassium
  • Method: Tetraphenylborate-turbidity
  • Case of 100
Specifications
Specifications
Photometry Tablet Reagents
Test kits, colorimeters and spectrophotometers
100 Tablets
Potassium T Tablets