Learn More
Lovibond™ Tintometer™ Phosphate (Ortho) 2 Reagent Ampoule, 0.05 to 5mg/L
For use with Lovibond test kits, colorimeters and spectrophotometers
Brand: Lovibond™ 380480
Additional Details : Weight : 0.30000kg Transport : UN number : 1760 Chem class : 8 Pack group : II
Description
- Range: 0.05 to 5mg/L
- Parameter: Ortho Phosphate
- Method: Stannous Chloride
- For use in test kits, colorimeters and spectrophotometers
- Case of 30
Specifications
Test kits, colorimeters and spectrophotometers | |
0.05 to 5mg/L | |
Phosphate (Ortho) 2 Reagent Ampoule |
30 | |
Stannous Chloride |