missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ Organo-Phosphonate No.2 Solution, 100mL

For use with Lovibond test kits, colorimeters and spectrophotometers

Brand:  Lovibond™ 465351

Additional Details : Weight : 0.30000kg

Product Code. 15332045

  • 421.00 DKK / 100mL
Estimated Shipment: 03-06-2024
to see stock.
Add to basket

Description

Description

  • Organo-Phosphonate No.2 Solution
  • 100mL
Specifications

Specifications

Test kits, colorimeters and spectrophotometers
100 mL
Organo-Phosphonate No.2 Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers