Learn More
Lovibond™ Tintometer™ Nitrite No. 1 Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand: Lovibond™ 505200
Additional Details : Weight : 0.09900kg
Description
- Nitrite No. 1 Tablets
- Bottle of 100
Specifications
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
100 Tablets | |
Nitrite No. 1 Tablets |