Learn More
Lovibond™ Tintometer™ Molybdate No. 1 HR / No. 2 HR Tablets Combi Pack
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand:  Lovibond™ 517631BT
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 15331915
  • 670.00 DKK / Pack of 100
Estimated Shipment: 09-07-2024
to see stock.
Add to basket
Description
Description
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
  • Range: 1 to 50mg/L
  • Parameter: Molybdate
  • Method: Thioglycolate
  • New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
  • With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
  • Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
  • Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
  • ‘BT’ is added to the end of the code to identify the new style of packaging
  • Simple, time-saving and practical
  • There are no safety risks if the tablets are used in line with the instructions supplied
  • Safety data sheets are available for all reagents
Specifications
Specifications
Photometry Tablet Reagents
250 Tablets
Molybdate No. 1 HR / No. 2 HR Tablets Combi Pack: 100 ea
Test kits, colorimeters and spectrophotometers
1 to 50mg/L