Learn More
Lovibond™ Tintometer™ Comparator 2000+ Disc 3/117, Iron Test, 1 to 10ppm
Glass color standards are fade-free, resistant to chemicals and scratchproof. Use these discs with the Comparator 2000+ for color comparator tests.
Brand: Lovibond™ 230110
This item is not returnable.
View return policy
Description
- The required accuracy of results is only ensured if stable, fade-free color standards are used
- Glass color standards are fade-free, resistant to chemicals and scratchproof
- Lovibond™ standards are made from colored glass filters
- Comply with international standards, e.g. ISO 7393/2
- Readings: 1, 2, 3, 4, 5, 6, 7, 8, 10mg/L
Specifications
Iron Test | |
Comparator 2000+ | |
1 to 10ppm | |
Comparator 2000+ Disc 3/117 |
Product Content Correction
Product Title
Lovibond™ Tintometer™ Comparator 2000+ Disc 3/117, Iron Test, 1 to 10ppm > Comparator 2000+ Disc 3/117
Spot an opportunity for improvement?Share a Content Correction