Learn More
Lovibond™ Tintometer™ CHECKIT™ Comparator D55 with Mirror Optics
Suitable for both mobile and static water analysis. Checkit™ Comparators are available with a number of different color scales and provide the basis for a comprehensive easy to to to use analysis system.
Brand: Lovibond™ 145010
Additional Details : Weight : 0.05900kg
Description
- The Lovibond™ Checkit™ comparator D55 with Mirror Optics is a compact, lightweight colorimetric unit
- Suitable for both mobile and static water analysis
- Available with a number of different color scales
- Provides the basis for a comprehensive easy to to to use analysis system
- Each Checkit™ Comparator Disc contains a continuous colour scale for exact color matching between standard and sample
- Each Checkit™ Comparator Kit is supplied in a carry case with comparator, vials, discs, reagents and method instructions for all tests
Specifications
Dia: 55mm | |
Checkit Comprator Tests | |
Checkit Comprator with Mirror Optics |
Water Quality Testing | |
Carry case with comparator, vials, discs, reagents and method instructions for all tests |