missing translation for 'onlineSavingsMsg'
Learn More

Tin solution for ICP/MS 10mg/L HNO3/HF matrix, Fisher chemical™

Catalog Number p-7083820
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Bottle
This item is not returnable. View return policy
7440-31-5
Sn
118.71
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
CHEBI:32990
tin
[Sn]
This item is not returnable. View return policy
7440-31-5
Sn
118.71
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
CHEBI:32990
tin
[Sn]

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 10mg/L
Packaging Bottle
Quantity 50 mL
Solvent or Matrix HNO3/HF
Chemical Name or Material Tin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'