missing translation for 'onlineSavingsMsg'
Få mere at vide

Timolol Hemihydrate, Mikromol™

Artikelnummer. 18082000
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18082000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18082000 Supplier Mikromol Supplier No. MM0440.240100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Timolol Hemihydrate
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Timolol Hemihydrate
CAS 91524-16-2
Analyt- eller komponentnavne Timolol Hemihydrate
Forsendelsestilstand Room Temperature
Molekylær formel 2 C13 H24 N4 O3 S . H2 O
Certificeringer/compliance ISO 17034
Synonym (2S)-1-[(1,1-Dimethylethyl)amino]-3-[[4-(4-morpholinyl)-1,2,5-thiadiazol-3-yl]oxy]-2-propanol Hydrate (2:1), 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-[[4-(4-morpholinyl)-1,2,5-thiadiazol-3-yl]oxy]-, hydrate (2:1), (S)-, 1,2,5-Thiadiazole, 2-propanol deriv., Betimol, Timolol Hemihydrate
InChI formel InChI=1S/2C13H24N4O3S.H2O/c2*1-13(2,3)14-8-10(18)9-20-12-11(15-21-16-12)17-4-6-19-7-5-17;/h2*10,14,18H,4-9H2,1-3H3;1H2/t2*10-;/m00./s1
SMIL O.CC(C)(C)NC[C@H](O)COc1nsnc1N2CCOCC2.CC(C)(C)NC[C@H](O)COc3nsnc3N4CCOCC4
IUPAC navn (2S)-1-(tert-butylamino)-3-[(4-morpholin-4-yl-1,2,5-thiadiazol-3-yl)oxy]propan-2-ol;hydrate
Molekylvægt (g/mol) 650.85
Mængde 100 mg
Formel vægt 650.3244 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.