Få mere at vide
Timiperone-d4 (Major)(Contain d0), TRC
Artikelnummer. 30450471
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30450471 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30450471 Leverandør Toronto Research Chemicals Leverandørnr. TRCT4431121MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Timiperone-d4
CAS 1185042-00-5
Molekylær formel C22H20D4FN3OS
Mængde 1 mg
Synonym 4-[4-(2,3-Dihydro-2-thioxo-1H-(benzimidazol-d4)-1-yl)-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone, 1-[1-[3-(4-Fluorobenzoyl)propyl]-4-piperidyl]-2-mercaptobenzimidazole-d4, 4-Fluoro-4-[4-(2-thioxo-1-benzimidazolinyl-d4)piperidino]butyrophenone, 1-[1-[3-(4-Fluorobenzoyl)propyl]-4-piperidyl]-2,3-(dihydrobenzimidazole-d4)-2-thione, Tolopelon
InChI formel InChI=1S/C22H24FN3OS/c23-17-9-7-16(8-10-17)21(27)6-3-13-25-14-11-18(12-15-25)26-20-5-2-1-4-19(20)24-22(26)28/h1-2,4-5,7-10,18H,3,6,11-15H2,(H,24,28)/i1D,2D,4D,5D
SMIL [2H]c1c([2H])c([2H])c2N(C3CCN(CCCC(=O)c4ccc(F)cc4)CC3)C(=S)Nc2c1[2H]
IUPAC navn 1-(4-fluorophenyl)-4-[4-(4,5,6,7-tetradeuterio-2-sulfanylidene-3H-benzimidazol-1-yl)piperidin-1-yl]butan-1-one
Molekylvægt (g/mol) 401.53
Formel vægt 401.1875