missing translation for 'onlineSavingsMsg'
Få mere at vide

TOKU-E™ Timentin ReadyMade™ Solution

Artikelnummer. 30044052
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 x 1 ml
Pakningsstørrelse:
10ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30044052 10 x 1 ml 10ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30044052 Leverandør TOKU-E™ Leverandørnr. T14710x1ML

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Timentin ReadyMade™ Solution is provided as a sterile-filtered solution of Timentin (Ticarcillin Disodium with Clavulanate Potassium 15:1) in water.

  • CAS Number: 116876-37-0
  • Molecular Formula: C15H14N2Na2O6S2 / C8H8KNO5
  • Molecular Weight: 665.64 g/mol (combined)
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Semi-synthetic
  • Spectrum: Timentin is broad-spectrum and has activity against Gram-negative cocci other than enterococci and methicillin-resistant staphylococci; Enterobacteriaceae, including drug-resistant strains; and anaerobes
  • Mechanism of Action: Timentin interferes with PBP (penicillin binding protein) activity involved in the final phase of peptidoglycan synthesis. PBPs are enzymes which catalyze a pentaglycine crosslink between alanine and lysine residues providing additional strength to the cell wall. Without a pentaglycine crosslink, the integrity of the cell wall is severely compromised and ultimately leads to cell lysis and death. Resistance to β-lactams is commonly due to cells containing plasmid encoded β-lactamases. Clavulanic acid is a 'suicide' or 'mechanism based' β-lactamase inhibitor. Clavulanic acid contains a β-lactam ring that binds strongly to β-lactamase at or near its activation site, thereby permanently inhibiting enzymatic activity. This action protects other beta-lactam antibiotics (penicillins, cephalosporins, etc.) from β-lactamase catalysis, thereby enhancing their antibacterial activity

Tekniske data

Kemisk navn eller materiale Ticarcillin Disodium with Clavulanate Potassium 15:1
Fysisk form Liquid
CAS 116876-37-0
Kilde Semi-synthetic
Emballage 2 mL plastic vial
Steril filtreret Yes, 0.22 μm
Sterilitet Sterile
Farve Colorless
Koncentration 95 to 105 mg/mL
Mængde 10 x 1 ml
Renhedsgrad noter >90% (HPLC)
Analyseprocentområde >75% Ticarcillin DiNa, >5% Clavulanate
Assay Procent Noter Timentin, Dried Basis
Molekylær formel C15H14N2Na2O6S2 / C8H8KNO5
Produktlinje ReadyMade™ Solutions
Forsendelsestilstand Ice Pack
Synonym Ticarcillin Disodium Salt/ Potassium Clavulanate mixture (15:1)
Molekylvægt (g/mol) 665.64 g/mol
Formel vægt 665.64 amu
Procent renhed >90%
Grad Antibiotic
Koncentration eller sammensætning (efter analyt eller komponenter) Timentin 10%
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.