Learn More
TIM-4 Rat anti-Mouse, Brilliant Violet 711, Clone: 21H12, BD Optibuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 742777
2945.71 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The 21H12 monoclonal antibody specifically binds to TIM-4. TIM-4 is encoded by Timd4 /(T cell immunoglobulin and mucin domain containing 4). TIM-4 is also known as Spleen, mucin-containing, knockout of lymphotoxin protein (SMUCKLER). TIM-4 is a single-pass type I membrane transmembrane glycoprotein belonging to the TIM family of the immunoglobulin superfamily. TIM-4 is expressed by macrophages and at low levels by dendritic cells. TIM-4 is a phosphatidylserine receptor that enhances the phagocytosis of apoptotic cells. It can also serve as a receptor for TIM-1, also known as the Hepatitis A virus cellular receptor 1 (Havcr1).
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
Specifications
TIM-4 | |
21H12 | |
Brilliant Violet 711 | |
TIMD-4, SMUCKLER, Tim4, Timd4 | |
Mouse TIM-4 Recombinant Protein | |
50 ÎĽg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 Îş |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.