missing translation for 'onlineSavingsMsg'
Få mere at vide

Tilorone Dihydrochloride, TRC

Artikelnummer. 30453113
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30453113 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453113 Supplier Toronto Research Chemicals Supplier No. TRCT440800100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Tilorone Dihydrochloride
CAS 27591-69-1
Molekylær formel C25 H34 N2 O3 . 2 Cl H
Mængde 100 mg
Synonym 9H-Fluoren-9-one','2,7-bis[2-(diethylamino)ethoxy]-','hydrochloride (1:2), 9H-Fluoren-9-one','2,7-bis[2-(diethylamino)ethoxy]-','dihydrochloride (9CI), Fluoren-9-one','2,7-bis[2-(diethylamino)ethoxy]-','dihydrochloride (8CI), Amixinum, Amyxin, DEAE-F, NSC 143969, RMI 10008DA, RMI 10028DA, Tilorone dihydrochloride, Tilorone hydrochloride
InChI formel InChI=1S/C25H34N2O3.2ClH/c1-5-26(6-2)13-15-29-19-9-11-21-22-12-10-20(30-16-14-27(7-3)8-4)18-24(22)25(28)23(21)17-19;;/h9-12,17-18H,5-8,13-16H2,1-4H3;2*1H
SMIL Cl.Cl.CCN(CC)CCOc1ccc2c(c1)C(=O)c3cc(OCCN(CC)CC)ccc23
IUPAC navn 2,7-bis[2-(diethylamino)ethoxy]fluoren-9-one;dihydrochloride
Molekylvægt (g/mol) 483.47
Formel vægt 482.2103
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.