missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tight Junction Protein 2 Antibody [FITC], Novus Biologicals Biologicals™
Shop alle Bio Techne produkterBeskrivelse
Tight Junction Protein 2 Polyclonal antibody specifically detects Tight Junction Protein 2 in Human,Mouse samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Tekniske data
Tekniske data
| Antigen | Tight Junction Protein 2 |
| Applikationer | ELISA, Immunoprecipitation, Western Blot |
| Klassifikation | Polyclonal |
| Konjugeret | FITC |
| Formulering | PBS |
| Gene Alias | C9DUPq21.11, DFNA51, DUP9q21.11, MGC26306, Tight junction protein 2, tight junction protein 2 (zona occludens 2), TJP2, X104, X104tight junction protein ZO-2, ZO2, ZO-2, ZO2Friedreich ataxia region gene X104 (tight junction protein ZO-2), Zona occludens protein 2, Zonula occludens protein 2 |
| Værtsarter | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 951-1190 of human Tight Junction Protein 2 (NP_004808.2).,, Sequence:, RSSEPVQHEESIRKPSPEPRAQMRRAASSDQLRDNSPPPAFKPEPPKAKTQNKEESYDFSKSYEYKSNPSAVAGNETPGASTKGYPPPVAAKPTFGRSILKPSTPIPPQEGEEVGESSEEQDNAPKSVLGKVKIFEKMDHKARLQRMQELQEAQNARIEIAQKHPDIYAVPIKTHKPDPGTPQHTSSRPPEPQKAPSRPYQDTRGSYGSDAEEEEYRQQLSEHSKRGYYGQSARYRDTEL |
| Oprensningsmetode | Affinity purified |
| Mængde | 0.1 mL |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?