Få mere at vide
Ticlopidine Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ticlopidine hydrochloride |
| CAS | 53885-35-1 |
| Analyt- eller komponentnavne | Ticlopidine hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C14 H14 Cl N S . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Ticlopidine Hydrochloride, 5-[(2-Chlorophenyl)methyl]-4,5,6,7-tetrahydrothieno[3,2-c]pyridine hydrochloride (1:1), 5-[(2-Chlorophenyl)methyl]-4,5,6,7-tetrahydrothieno[3,2-c]pyridine hydrochloride, 4C32, 53-32C, Anagregal, Caudaline, Nichistate, Panaldine, Panapidin, Ticlodone, Ticlodox, Ticlopidine hydrochloride, Ticlosin, Tiklid |
| InChI formel | InChI=1S/C14H14ClNS.ClH/c15-13-4-2-1-3-11(13)9-16-7-5-14-12(10-16)6-8-17-14;/h1-4,6,8H,5,7,9-10H2;1H |
| Løsningstype | Neat |
| SMIL | Cl.Clc1ccccc1CN2CCc3sccc3C2 |
| Vis mere |