Få mere at vide
Ticlopidine EP Impurity J (as Dihydrochloride), Mikromol™
Artikelnummer. 30458021
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30458021 4 x 25 mg
30049310 25 mg
30051805 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30458021 Leverandør Mikromol Leverandørnr. MM0150.1000254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ticlopidine EP Impurity J (as Dihydrochloride), Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N'-Bis(2-chlorobenzyl)ethane-1,2-diamine Dihydrochloride
CAS 2056-18-0
Analyt- eller komponentnavne N,N'-Bis(2-chlorobenzyl)ethane-1,2-diamine Dihydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C16 H18 Cl2 N2 . 2 Cl H
Certificeringer/compliance ISO 17025
Synonym N,N'-Bis(2-chlorobenzyl)ethane-1,2-diamine Dihydrochloride, Ticlopidine Hydrochloride Imp. J (EP) as Dihydrochloride, Ticlopidine Hydrochloride Imp. J (EP), Ticlopidine Imp. J (EP), Ticlopidine Hydrochloride Impurity J as Dihydrochloride, Ticlopidine Impurity J as Dihydrochloride
InChI formel InChI=1S/C16H18Cl2N2.2ClH/c17-15-7-3-1-5-13(15)11-19-9-10-20-12-14-6-2-4-8-16(14)18;;/h1-8,19-20H,9-12H2;2*1H
Løsningstype Neat
SMIL Cl.Cl.Clc1ccccc1CNCCNCc2ccccc2Cl
IUPAC navn N,N'-bis[(2-chlorophenyl)methyl]ethane-1,2-diamine;dihydrochloride
Molekylvægt (g/mol) 382.16
Mængde 4 x 25 mg
Formel vægt 380.0381 g/mol