Learn More
Tiaprofenic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Tiaprofenic acid |
| CAS | 33005-95-7 |
| Molecular Formula | C14 H12 O3 S |
| Quantity | 250 mg |
| Synonym | Tiaprofenic acid, (2RS)-2-(5-Benzoylthiophen-2-yl)propanoic acid |
| InChI Formula | InChI=1S/C14H12O3S/c1-9(14(16)17)11-7-8-12(18-11)13(15)10-5-3-2-4-6-10/h2-9H,1H3,(H,16,17) |
| SMILES | CC(C(=O)O)c1ccc(s1)C(=O)c2ccccc2 |
| IUPAC Name | 2-(5-benzoylthiophen-2-yl)propanoic acid |
| Molecular Weight (g/mol) | 260.31 |
| Formula Weight | 260.0507 |