Få mere at vide
Tianeptine Ethyl Ester, TRC
Artikelnummer. 30451222
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30451222 10 mg
30453999 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30451222 Leverandør Toronto Research Chemicals Leverandørnr. TRCT43683010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 66981-77-9
Molekylær formel C23H29ClN2O4S
Molekylvægt (g/mol) 465.01
Synonym 7-[(3-Chloro-6,11-dihydro-6-methyl-5,5-dioxidodibenzo[c,f][1,2]thiazepin-11-yl)amino]-heptanoic Acid Ethyl Ester, 7-[(3-chloro-6,11-dihydro-6-methyldibenzo[c,f][1,2]thiazepin-11-yl)amino]-heptanoic Acid Ethyl Ester S,S-Dioxide, Dibenzo[c,f][1,2]thiazepine, Heptanoic Acid Derivative, Tianeptine Impurity B
IUPAC navn ethyl 7-[(3-chloro-6-methyl-5,5-dioxo-11H-benzo[c][1,2]benzothiazepin-11-yl)amino]heptanoate
SMIL CCOC(=O)CCCCCCNC1c2ccccc2N(C)S(=O)(=O)c3cc(Cl)ccc13
Tekniske data
Kemisk navn eller materiale Tianeptine Ethyl Ester
Mængde 10 mg
InChI formel InChI=1S/C23H29ClN2O4S/c1-3-30-22(27)12-6-4-5-9-15-25-23-18-10-7-8-11-20(18)26(2)31(28,29)21-16-17(24)13-14-19(21)23/h7-8,10-11,13-14,16,23,25H,3-6,9,12,15H2,1-2H3
Formel vægt 464.1537