Få mere at vide
Tianeptine-d12, TRC
Artikelnummer. 30453607
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30453607 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453607 Leverandør Toronto Research Chemicals Leverandørnr. TRCT4368025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tianeptine-d12
CAS 1189870-26-5
Molekylær formel C21H13D12ClN2O4S
Mængde 5 mg
Synonym Tianeptine D12
InChI formel InChI=1S/C21H25ClN2O4S/c1-24-18-9-6-5-8-16(18)21(23-13-7-3-2-4-10-20(25)26)17-12-11-15(22)14-19(17)29(24,27)28/h5-6,8-9,11-12,14,21,23H,2-4,7,10,13H2,1H3,(H,25,26)/i2D2,3D2,4D2,7D2,10D2,13D2
SMIL [2H]C([2H])(NC1c2ccccc2N(C)S(=O)(=O)c3cc(Cl)ccc13)C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O
IUPAC navn 7-[(3-chloro-6-methyl-5,5-dioxo-11H-benzo[c][1,2]benzothiazepin-11-yl)amino]-2,2,3,3,4,4,5,5,6,6,7,7-dodecadeuterioheptanoic acid
Molekylvægt (g/mol) 449.03
Formel vægt 448.1977