Få mere at vide
Tiamulin Hydrogen Fumarate, LoGiCal
Artikelnummer. 30462061
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30462061 10 mg
1 options
Artikelnummer. 30462061 Leverandør LoGiCal Leverandørnr. LGCFOR0394.00
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tiamulin fumarate
CAS 55297-96-6
Analyt- eller komponentnavne Tiamulin fumarate
Forsendelsestilstand Room Temperature
Molekylær formel C28 H47 N O4 S . C4 H4 O4
Certificeringer/compliance ISO 9001
Synonym Tiamulin Hydrogen Fumarate, [[2-(Diethylamino)ethyl]thio]acetic acid (3aS,4R,5S,6S,8R,9S,9aR,10R)-6-ethenyldecahydro-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-3a,9-propano-3aH-cyclopentacycloocten-8-yl ester (2E)-2-butenedioate (1:1) (salt), 3a,9-Propano-3aH-cyclopentacyclooctene, acetic acid deriv., 14-Deoxy-14-[(2-diethylaminoethyl)-mercaptoacetoxy]mutilin hydrogen fumarate, 81723hfu, Dynamutilin, SQ 22947, Sandoz 81723hfu, Thiamutilin fumarate, Tiamulin fumarate, Tiamulin hydrogen fumarate, Tiamutin, Tiavet, [[2-(Diethylamino)ethyl]thio]acetic acid [3aS-(3aalpha,4beta,5alpha,6alpha,8beta,9alpha,9abeta,10S*)]-6-ethenyldecahydro-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-3a,9-propano-3aH-cyclopentacycloocten-8-yl ester (E)-2-butenedioate (1:1) (salt)
InChI formel InChI=1S/C28H47NO4S.C4H4O4/c1-8-26(6)17-22(33-23(31)18-34-16-15-29(9-2)10-3)27(7)19(4)11-13-28(20(5)25(26)32)14-12-21(30)24(27)28;5-3(6)1-2-4(7)8/h8,19-20,22,24-25,32H,1,9-18H2,2-7H3;1-2H,(H,5,6)(H,7,8)/b;2-1+/t19-,20+,22-,24+,25+,26-,27+,28+;/m1./s1
Løsningstype Neat
SMIL CCN(CC)CCSCC(=O)O[C@@H]1C[C@@](C)(C=C)[C@@H](O)[C@H](C)[C@@]23CC[C@@H](C)[C@]1(C)[C@@H]2C(=O)CC3.OC(=O)\C=C\C(=O)O
IUPAC navn (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5-hydroxy- 4,6,9,10-tetramethyl-1-oxodecahydro-3a,9-propano-3aH-cyclopentacycloocten-8-yl [[2-(diethylamino)ethyl]sulfanyl]acetate monofumarate
Industry Type Clinical Testing
Molekylvægt (g/mol) 609.81
Mængde 10 mg
Formel vægt 609.3335 g/mol