missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIAL1 Antibody (6E3), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP3-11834UV
Additional Details : Weight : 0.00970kg
Description
TIAL1 Monoclonal antibody specifically detects TIAL1 in Human, Mouse, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), FlowSpecifications
TIAL1 | |
Monoclonal | |
DyLight 350 | |
aging-associated gene 7 protein, MGC33401, TCBP, T-cluster binding protein, TIA1 cytotoxic granule-associated RNA binding protein-like 1, TIA1 cytotoxic granule-associated RNA-binding protein-like 1, TIA-1 related protein, TIA-1-related nucleolysin, TIA-1-related protein, TIARnucleolysin TIAR | |
GST-tagged recombinant human TIAL1 | |
0.1 mL | |
Apoptosis | |
7073 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry | |
6000 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Primate | |
Purified |