Få mere at vide
THZ531, MedChemExpress
Artikelnummer. 16422886
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
Pakningsstørrelse:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16422886 25 mg 25mg
16403878 10 mg -
16456597 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16422886 Leverandør Medchem Express Leverandørnr. HY103618
MedChemExpress THZ531 is a selective and covalent inhibitor of both CDK12 and CDK13 with IC50s of 158 nM and 69 nM, respectively.
THZ531 is a selective and covalent inhibitor of both CDK12 and CDK13 with IC50s of 158 nM and 69 nM, respectively.
Tekniske data
Kemisk navn eller materiale THZ531
CAS 1702809-17-3
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C30H32ClN7O2
Mængde 25 mg
Opløselighedsinformation DMSO : 14.29 mg/mL (25.61 mM; Need ultrasonic)
SMIL ClC(C(C1=CNC2=CC=CC=C12)=N3)=CN=C3N[C@@H]4CCCN(C(C5=CC=C(NC(/C=C/CN(C)C)=O)C=C5)=O)C4
Molekylvægt (g/mol) 558.07
Formel vægt 558.07
Procent renhed 98.11%
Grad Research
Fysisk form Powder
Til brug med (applikation) Cancer-Kinase/protease