missing translation for 'onlineSavingsMsg'
Få mere at vide

Thymol Blue, Supelco™

Artikelnummer. 15897650
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
15897650 25 g
15887650 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15897650 Leverandør Supelco Leverandørnr. 1081760025
 Begrænset vare
Forventet forsendelse: 14-12-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Indicator ACS, Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 76-61-9

Specifications

Kemisk navn eller materiale Thymol Blue
Smeltepunkt 221°C (decomposition)
MDL nummer MFCD00005869
Synonym Thymol blue, Thymolsulfonphthalein, Thymol blue
Mængde 25 g
Molekylvægt (g/mol) 466.59
Grad Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.