Få mere at vide
Thymol-b-D-glucopyranoside, TRC
Artikelnummer. 30450253
Skift visning
Mængde:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30450253 50 mg
30451018 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30450253 Leverandør Toronto Research Chemicals Leverandørnr. TRCT41309050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Thymol-b-D-glucopyranoside
CAS 20772-23-0
Molekylær formel C16H24O6
Mængde 50 mg
InChI formel InChI=1S/C16H24O6/c1-8(2)10-5-4-9(3)6-11(10)21-16-15(20)14(19)13(18)12(7-17)22-16/h4-6,8,12-20H,7H2,1-3H3/t12-,13-,14+,15-,16-/m1/s1
SMIL CC(C)c1ccc(C)cc1O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O
IUPAC navn (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(5-methyl-2-propan-2-ylphenoxy)oxane-3,4,5-triol
Molekylvægt (g/mol) 312.36
Formel vægt 312.1573