Få mere at vide
Thymine-d3, TRC
Artikelnummer. 30452591
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30452591 10 mg
30451403 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452591 Leverandør Toronto Research Chemicals Leverandørnr. TRCT41215210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 68941-98-0
Molekylær formel C5 2H3 H3 N2 O2
Molekylvægt (g/mol) 129.13
Synonym 2,4(1H,3H)-Pyrimidinedione, 5-(methyl-d3)- (9CI), Thymine-d3, 5-(Methyl-d3)-2,4(1H,3H)-pyrimidinedione, Thymine-D3 (methyl-D3), Thymine-α,α,α-d3
IUPAC navn 5-(trideuteriomethyl)-1H-pyrimidine-2,4-dione
SMIL [2H]C([2H])([2H])C1=CNC(=O)NC1=O
Tekniske data
Kemisk navn eller materiale Thymine, methyl-D3
Mængde 10 mg
InChI formel InChI=1S/C5H6N2O2/c1-3-2-6-5(9)7-4(3)8/h2H,1H3,(H2,6,7,8,9)/i1D3
Formel vægt 129.0618