Få mere at vide
Thymine-alpha,alpha,alpha,6-d4 Glycol (mixture of diastereomers), CDN
Artikelnummer. 30320054
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320054 0.01 g
30319639 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320054 Leverandør CDN Leverandørnr. CDND43500.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Thymine Glycol D4 (6-D,methyl-D3)
CAS 132523-94-5
Molekylær formel C5 D4 H4 N2 O4
Mængde 0.01 g
Synonym 2,4(1 H,3 H)-Pyrimidinedione-6-d, dihydro-5,6-dihydroxy-5-(methyl-d3)- (9 CI), Dihydro-5,6-dihydroxy-5-(methyl-d3)-2,4(1 H,3 H)-pyrimidinedione-6-d, 5,6-Dihydroxy-5,6-dihydrothymine-α,α,α,6-d4, Thymine glycol-d4, Thymine-α,α,α,6-d4 glycol, Thymine glycol D4 (6-D,methyl-D3)
InChI formel InChI=1 S/C5H8N2O4/c1-5(11)2(8)6-4(10)7-3(5)9/h2,8,11 H,1H3,(H2,6,7,9,10)/i1D3,2 D
SMIL [2 H]C([2 H])([2 H])C1(O)C(=O)NC(=O)NC1([2 H])O
IUPAC navn 6-deuterio-5,6-dihydroxy-5-(trideuteriomethyl)-1,3-diazinane-2,4-dione
Molekylvægt (g/mol) 164.153
Formel vægt 164.074 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity